Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p53 (p Thr55) Rabbit anti-Human, Clone: SC58-01, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267493
Description
p53 (p Thr55) Monoclonal antibody specifically detects p53 (p Thr55) in Human samples. It is validated for Western Blot.Specifications
p53 (p Thr55) | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:100-1:500 | |
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
Rabbit | |
Protein A purified | |
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
7157 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
SC58-01 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
TP53 | |
Synthetic phospho-peptide corresponding to residues surrounding Thr55 of Human p53 aa 31-80/ 393. (SwissProt: P04637 Human) | |
100 μL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction