Learn More
p53 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259153
Catalog No. NBP259153
Add to cart
Description
Description
p53 Polyclonal antibody specifically detects p53 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications
Specifications
p53
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Rabbit
IgG
50 ug
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary
7157
Western Blot, ChIP assay
Western Blot 1:200, Chromatin Immunoprecipitation 5 ug
Purified
TP53
The exact immunogen is propietary information.
Protein G purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human