Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p53, Rabbit Monoclonal Antibody (Clone Y5)

Employ part of a comprehensive antibody portfolio with the Thermo Scientific™ p53, Rabbit Monoclonal Antibody (Clone Y5).

$254.50 - $1115.00

Specifications

Antigen p53
Clone Y5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RM2103R7
View Documents
Thermo Scientific™
RM2103R7
Ready-to-use for IHC (UVLP) 7mL
Each for $749.00
Add to cart
 
RM2103RQ
View Documents
Thermo Scientific™
RM2103RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $1,013.00
Add to cart
 
RM2103S
View Documents
Thermo Scientific™
RM2103S
Culture Supernatant with Azide 1mL
Each for $1,115.00
Add to cart
 
RM2103S0
View Documents
Thermo Scientific™
RM2103S0
Culture Supernatant with Azide 100μL
Each for $254.50
Add to cart
 
RM2103R1
View Documents
Thermo Scientific™
RM2103R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Employ part of a comprehensive antibody portfolio with the Thermo Scientific™ p53, Rabbit Monoclonal Antibody (Clone Y5). Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein. p53 senses DNA damage and possibly facilitaes repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma. p53 is detected by immunostains only if the mutation causes protein stability (although protein may be nonfunctional) or a physiologic response. The physiologic response may be due to genetic changes induced by cellular stress from anoxia or aneuploidy; or genetic damage to other pathways controlling p53 such as MDM2 or alternative reading frame.

Host Species: Rabbit

Clone: Y5

Species Reactivity: Human.Others not known.

Immunogen: A synthetic peptide derived from N- terminal residues of human p53

Molecular Weight: 53kDa

Positive Control: Colon or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

p53
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
Y5
Monoclonal
Rabbit
Cancer and Tumor Biology
A synthetic peptide derived from N- terminal residues of human p53
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit