Learn More
Invitrogen™ p53 (Total/Phospho) Human InstantOne™ ELISA Kit Non-distribution product as customer accommodation.
InstantOne ELISA
Manufacturer:  Invitrogen™ 858612311
Catalog No. 50-246-267
Add to cart
Alert:
The Total/Phospho InstantOne ELISAs contain reagents for a total of 96 wells (24 wells total/72 wells phospho)
Please contact Technical Service for a lot specific COA
Includes: User Manual
InstantOne™ Assay Plate - 96 well (sealed)
Cell Lysis Buffer (5X)
Enhancer Solution
Wash Buffer (10X)
p53 (Total) Capture Antibody Reagent
phospho-p53 (Ser15) Capture Antibody Reagent
p53 (Total) Detection Antibody Reagent
phospho-p53 (Ser15) Detection Antibody Reagent
Positive Control Cell Lysate (lyophilized)
Detection Reagent (TMB Substrate)
Stop Solution
Plate Seal
Description
Description
The InstantOne™ ELISA is specifically engineered for accurate measurement of total and phosphorylated human p53 in cell lysates. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
The tumor suppressor protein, p53, is a sequence specific transcription factor that is activated by cellular stress. p53 mediates cell cycle arrest or apoptosis in response to DNA damage or starvation for pyrimidine nucleotides. p53 is up-regulated in response to stress signals and stimulated to activate transcription of specific genes, resulting in expression of p21waf1 and other proteins involved in G1 or G2/M arrest. The structure of p53 comprises an N-terminal transactivation domain, a central DNA-binding domain, an oligomerisation domain, and a C-terminal regulatory domain. There are various phosphorylation sites on p53, of which the phosphorylation at Ser15 is important for p53 activation and stabilization. p53 has been characterized to play a role in blocking the proliferative action of damaged cells and act as an anticancer agent. Phosphorylation of Ser392 in p53 has been shown to associate with the formation of human tumors. In addition, p53 has also been linked to the effects of aging and oxidative stress and an increase in p53 has been linked to deficits in LTP (Long Term Potentiation) in learning and memory. p53 is found in very low levels in normal cells, however, in a variety of transformed cell lines, it is expressed in high amounts, and believed to contribute to transformation and malignancy. Mutants of p53 that frequently occur in a number of different human cancers fail to bind the consensus DNA binding site, and cause the loss of tumor suppressor activity. Alterations of the TP53 gene occur not only as somatic mutations in human malignancies, but also as germline mutations in some cancer-prone families such as Li-Fraumeni syndrome.
Specifications
Specifications
P04637
ELISA
Capture Antibody
RUO
2°C to 8°C
1 hr. 10 min.
BCC7,LFS1,P53,TRP53
96 Tests
Cell Lysates
Colorimetric Microplate Reader
Biotin
7157
ELISA Kit
Cell Lysate,20 μL
1 hr. 20 min.
Human
TP53
Human
HRP