Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p57 Kip2 Mouse anti-Human, Clone: 1F4A9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Thermo Scientific MA538493

Encompass

Catalog No. PIMA538493


Add to cart

Description

Description

This antibody has been tested in indirect ELISA.

p57 Kip2 Monoclonal antibody specifically detects p57 Kip2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

p57 Kip2
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P49918
AL024410; BWCR; BWS; CDKI; CDKN1C; cyclin dependent kinase inhibitor 1C; cyclin-dependent kinase inhibitor 1C; cyclin-dependent kinase inhibitor 1C (P57); cyclin-dependent kinase inhibitor 1C (p57, Kip2); cyclin-dependent kinase inhibitor p57; IMAGE; KIP2; p57; p57 Kip2; p57(kip2); p57Kip2; p57KIP2 A3sh; WBS
Purified recombinant fragment of human CDKN1C expressed in E. Coli.
Protein G
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1F4A9
Unconjugated
CDKN1C
Liquid, Purified
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1028
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit