Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p62/SQSTM1 Rabbit anti-Human, Mouse, Canine, DyLight 755, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149954IR

Catalog No. NBP149954IR


Add to cart

Description

Description

p62/SQSTM1 Polyclonal antibody specifically detects p62/SQSTM1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

p62/SQSTM1
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
Rabbit
IgG
100 ul
Apoptosis, Autophagy, Cancer, Macroautophagy, mTOR Pathway, Neurodegeneration, Protein Turnover, Regulatory Immunology, Signal Transduction, Stem Cell Signaling Pathway, Ubiquitin Proteasome Pathway
Primary
8878
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified
SQSTM1
The immunogen for this product maps to a region between residue 325 and 375 of human Sequestosome-1 using the numbering given in entry NP_003891.1 (GeneID 8878).
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Canine, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only