Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p67phox/NOXA2 Antibody (001), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290316APC
Description
p67phox/NOXA2 Monoclonal antibody specifically detects p67phox/NOXA2 in Human samples. It is validated for ELISASpecifications
p67phox/NOXA2 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
001 | |
ELISA | |
autosomal 2, NADPH oxidase activator 2, NCF-2, neutrophil cytosol factor 2, neutrophil cytosolic factor 2, neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal2), NOXA2FLJ93058, p67phox, p67-phox | |
Rabbit immunized with purified, recombinant Human p67phox/NOXA2 (Accession#: AAH01606.1, Met1-Val526) | |
0.1 mL | |
Cancer, Endocrinology, Immunology | |
4688 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction