Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73 Mouse anti-Human, Clone: P73/2531, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen p73
Clone P73/2531
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122948
View Documents
Novus Biologicals
NBP30773720UG
20 μg
Each for $138.37
Add to cart
 
NB122947
View Documents
Novus Biologicals
NBP30773710UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

p73 Monoclonal antibody specifically detects p73 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

p73
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Recombinant full-length human p73 protein
Monoclonal
Protein A or G purified
P73/2531
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
10 mM PBS with 0.05% BSA
7161
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit