Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73, Rabbit anti-Human, Alexa Fluor 700, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056435AF700

Catalog No. NB003543


Add to cart

Description

Description

p73 Polyclonal specifically detects p73 in Human samples. It is validated for Western Blot.
Specifications

Specifications

p73
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Rabbit
IgG
0.1 mL
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary
This peptide sequence is 100% conserved across multiple p73 isoforms and between rat, mouse, dog and bovine p73. Researchers are encourage to use the the immunogen sequence information and available Bioinformatics data bases to obtain additional information regarding the amino acid sizes associated with various p73 isoforms.
Western Blot
Western Blot
Affinity Purified, Conjugated
TP73
A synthetic peptide CTHSPYAQPSS corresponding to amino acids 95-104 of human p73 was used as immunogen.
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit