Learn More
P7C3, Tocris™
Neuroprotective compound
Manufacturer:  R&D Systems 4076/50
Quantity 50mg
 View more versions of this product
Catalog No. 40-765-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
474.19
P7C3
C21H18Br2N2O
3, 6-Dibromo-α-[(phenylamino)methyl]-9H-carbazole-9-ethanol
Specifications
50mg
>99%
474.19
P7C3
301353-96-8
C21H18Br2N2O
3, 6-Dibromo-α-[(phenylamino)methyl]-9H-carbazole-9-ethanol
Safety and Handling
Recommended Storage : Store at Room Temperature