Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA28 Activator beta Subunit/PSME2 Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP300350C

Catalog No. NB065700


Add to cart

Description

Description

PA28 Activator beta Subunit/PSME2 Polyclonal antibody specifically detects PA28 Activator beta Subunit/PSME2 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

PA28 Activator beta Subunit/PSME2
DyLight 650
50mM Sodium Borate
11S regulator complex beta subunit, 11S regulator complex subunit beta, Activator of multicatalytic protease subunit 2, MCP activator, 31-kD subunit, PA28b, PA28betacell migration-inducing protein 22, proteasome (prosome, macropain) activator subunit 2 (PA28 beta), Proteasome activator 28 subunit beta, proteasome activator 28-beta, proteasome activator complex subunit 2, proteasome activator hPA28 subunit beta, REGbeta, REG-beta
Produced in rabbits immunized with purified, recombinant PA28 Activator beta Subunit/PSME2 (Q9UL46; Met1-Tyr239).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Neuroscience, Ubiquitin Proteasome Pathway
Primary
5721
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit