Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA28 Activator gamma Subunit/PSME3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen PA28 Activator gamma Subunit/PSME3
Applications KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094880
View Documents
Novus Biologicals
NBP303887100UL
100 μg Each for $320.71
Add to cart
 
NB094881
View Documents
Novus Biologicals
NBP30388720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

PA28 Activator gamma Subunit/PSME3 Polyclonal antibody specifically detects PA28 Activator gamma Subunit/PSME3 in Human, Mouse samples. It is validated for Western Blot, KnockDown
Specifications

Specifications

PA28 Activator gamma Subunit/PSME3
Unconjugated
Rabbit
Core ESC Like Genes, Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway
PBS with 50% glycerol, pH7.3.
10197
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
KnockDown, Western Blot
Purified
RUO
Human, Mouse
11S regulator complex gamma subunit, 11S regulator complex subunit gamma, Activator of multicatalytic protease subunit 3, Ki, Ki antigen, PA28g, PA28gamma, PA28-gamma, PA28GKi nuclear autoantigen, proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki), Proteasome activator 28 subunit gamma, proteasome activator 28-gamma, proteasome activator complex subunit 3, REG-gamma
A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human PA28 Activator gamma Subunit/PSME3 (NP_005780.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit