Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAAF1 Mouse anti-Human, Mouse, FITC, Clone: OTI1F11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273214F
Description
PAAF1 Monoclonal specifically detects PAAF1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PAAF1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PAAF1 | |
Full length recombinant protein of human PAAF1 (NP_079431) produced in HEK293T cells. | |
0.1 mL | |
80227.0 | |
Store at 4°C in the dark. |
Immunocytochemistry, Immunofluorescence | |
OTI1F11 | |
Immunocytochemistry/Immunofluorescence | |
FLJ11848, PAAF, proteasomal ATPase-associated factor 1, Protein G-16, Rpn14, WD repeat domain 71, WD repeat-containing protein 71, WDR71 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title