Learn More
PAAF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 176501AP
Catalog No. 50-173-1703
Add to cart
Description
Description
PAAF1 inhibits proteasome 26S assembly and proteolytic activity by impairing the association of the 19S regulatory complex with the 20S core. In case of HIV-1 infection, recruited by viral Tat to the HIV-1 promoter, where it promotes the recruitment of 19S regulatory complex through dissociation of the proteasome 26S. This presumably promotes provirus transcription efficiency.
Specifications
Specifications
PAAF1
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BRP4
PAAF, PAAF1, Protein G 16, Rpn14, WDR71
Rabbit
IgG
150 μL
-20°C
Primary
80227
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PAAF1
Liquid
PAAF1
PAAF1 Fusion Protein Ag11892
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse