Learn More
Cell Biolabs IncSupplier Diversity Partner PAAV-RC1 VECTOR  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK421
The pAAV-RC1 vector contains the rep and cap genes required to generated recombinant AAV of serotype 1. Co-transfect with other packaging plasmids and an expression vector into 293 cells for AAV-1 packaging.
Catalog No. 50-673-895