Learn More
Cell Biolabs IncSupplier Diversity Partner PAAV-RC5 VECTOR  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK425
The pAAV-RC5 vector contains the rep and cap genes required to generated recombinant AAV of serotype 5. Co-transfect with other packaging plasmids and an expression vector into 293 cells for AAV-5 packaging.
Catalog No. 50-673-899