Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PABPN1L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548309

Catalog No. PA548309

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with xenopus, mouse and rat based on sequence homology.

Binds the poly(A) tail of mRNA.
Specifications

Specifications

PABPN1L
Polyclonal
PBS with 0.09% sodium azide
A6NDY0
Embryonic polyadenylate-binding protein 2, Embryonic poly(A)-binding protein 2, ePABP-2, ePABP2, Embryonic poly(A)-binding protein type II, Poly(A)-binding protein nuclear-like 1, PABPN1L, EPABP2, PABPNL1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
390748
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PABPN1L
Liquid
PABPN1L
KLH conjugated synthetic peptide between 166-194 amino acids from the Central region of human PABPN1L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit