Learn More
Alfa Aesar™ PACAP (6-27), Human, Ovine, Rat
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66872EXD
 View more versions of this product
Catalog No. AAJ66872EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2638.13
Human, Ovine, Rat
Solid
RUO
PACAP (6-27), human, ovine, rat
HPLC
0.5mg
Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2
-30° to -10°C