Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACAP/ADCYAP1 Sheep anti-Human, Alexa Fluor 700, Polyclonal, Novus Biologicals™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Sheep Polyclonal Antibody
Supplier: Novus Biologicals NB10062241AF700
Description
PACAP/ADCYAP1 Polyclonal antibody specifically detects PACAP/ADCYAP1 in Human samples. It is validated for ELISASpecifications
PACAP/ADCYAP1 | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 700 | |
adenylate cyclase activating polypeptide 1 (pituitary), PACAPMGC126852, pituitary adenylate cyclase-activating polypeptide | |
Proprietary, 14 amino acid peptide sequence of PACAP, conjugated to KLH. | |
0.1 mL | |
Signal Transduction | |
116 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction