Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PACAP Receptor Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533318
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset, Bovine (100%); Dog, Pig (94%); Mouse, Rat, Elephant, Panda (89%). Antigen retrieval requires Proteinase K treatment.
This gene encodes type I adenylate cyclase activating polypeptide receptor, which is a membrane-associated protein and shares significant homology with members of the glucagon/secretin receptor family. This receptor mediates diverse biological actions of adenylate cyclase activating polypeptide 1 and is positively coupled to adenylate cyclase. Alternative splicing of two exons of this gene generates four major splice variants, but their full-length nature has not been determined. PACAP receptor type 1 expression has been documented in adipose, adrenal, bone, brain, colon, ganglion, heart, lung, ovary, pancreas, placenta, spleen, and uterus. ESTs have been isolated from brain and nerve libraries.
Specifications
PACAP Receptor | |
Polyclonal | |
Unconjugated | |
ADCYAP1R1 | |
2900024I10Rik; ADCYAP receptor type I; ADCYAP1 R1; ADCYAP1R 1; Adcyap1r1; adenylate cyclase activating polypeptide 1 (pituitary) receptor type I; adenylate cyclase activating polypeptide 1 receptor 1; adenylate cyclase activating polypeptide 1 type 1 receptor; AI846590; alternatively spliced product of PACAP receptor; G-protein coupled receptor; PAC 1; PAC1; PAC1 receptor; Pac1 receptors; PAC1R; PAC1-R; PACAP R 1; PACAP Receptor; PACAP receptor 1; PACAP type I receptor; PACAP type IA receptor; PACAP/VIP receptors; PACAP1 R; PACAP1 receptor; PACAP1-R; PACAPR; PACAPR1; PACAP-R1; PACAP-R-1; PACAPR1A; PACAP-R1A; PACAP-R-1A; PACAPRI; pituitary adenylate cyclase activating polypeptide 1 receptor; pituitary adenylate cyclase activating polypeptide 1 receptor (1); pituitary adenylate cyclase activating polypeptide 1 receptor type I Hiphop; pituitary adenylate cyclase activating polypeptide receptor; pituitary adenylate cyclase-activating polypeptide type 1 receptor; pituitary adenylate cyclase-activating polypeptide type 1 receptor hop 1 splice variant; pituitary adenylate cyclase-activating polypeptide type I receptor; pituitary adenylate cyclase-activating polypeptide type IA receptor; pvr1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117, 319095 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P41586, Q29627 | |
ADCYAP1R1 | |
Synthetic 18 amino acid peptide from N-terminal extracellular domain of human ADCYAP1R1. | |
50 μg | |
Primary | |
Human, Monkey, Bovine | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction