Learn More
Alfa Aesar™ PACAP-Related Peptide (1-29), Rat
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66839EXD
 View more versions of this product
Catalog No. AAJ66839EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
3361.86
Rat
Solid
RUO
PRP
HPLC
0.5mg
Asp-Val-Ala-His-Glu-Ile-Leu-Asn-Glu-Ala-Tyr-Arg-Lys-Val-Leu-Asp-Gln-Leu-Ser-Ala-Arg-Lys-Tyr-Leu-Gln-Ser-Met-Val-Ala
-30° to -10°C
PACAP-Related Peptide (1-29), rat