Learn More
Alfa Aesar™ PACAP-Related Peptide, Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66104MCR
 View more versions of this product
Catalog No. AAJ66104MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
3146.60
Human
Solid
RUO
Pituitary adenylate cyclase activating polypeptide-related peptide
HPLC
1mg
Asp-Val-Ala-His-Gly-Ile-Leu-Asn-Glu-Ala-Tyr-Arg-Lys-Val-Leu-Asp-Gln-Leu-Ser-Ala-Gly-Lys-His-Leu-Gln-Ser-Leu-Val-Ala
-30° to -10°C
PACAP-Related Peptide, human