Learn More
Thermo Scientific™ Pacific Hemostasis™ Factor Assays
Detect and quantify factor deficiencies in patient plasmas with these factor assays.
 View more versions of this product
Catalog No. 22-176446
  Check Availability
Add to cart
Specifications
Description
Detect and quantify factor deficiencies in patient plasmas with Thermo Scientific™ Pacific Hemostasis™ Factor Assays.
Pacific Hemostasis Factor Assays include a complete line of lyophilized congenital factor deficient plasmas for performing factor assays.
  • All factor deficient plasmas are prepared from congenitally deficient patient donors or immunodepleted plasma, with the exception of Factor II.
  • Factor II Deficient Plasma is a mix of human and bovine plasma.
  • Plasmas are packaged 10 x 1mL; Diluting Fluid is packaged 2 x 100mL.
Description & Specifications
Specifications
Factor V Deficient Plasma