Learn More
Thermo Scientific™ Pacific Hemostasis™ FDP Assay
Fisherbrand™ Pacific Hemostasis FDP Assay is a fast, easy-to-use latex agglutination assay for use in measuring fibrinogen degradation products (FDP) in serum and urine.
 View more versions of this product
Catalog No. 22-249990
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is not returnable. View return policy
Description
Description
Measure fibrinogen degradation products (FDP) in serum and urine with Thermo Scientific™ Pacific Hemostasis™ FDP Assay, a fast, easy-to-use latex agglutination assay used in the diagnosis of disseminated intravascular coagulation (DIC), deep vein thrombosis (DVT), pulmonary embolism (PE), certain pregnancy disorders and myocardial infarction.
Features:
  • Latex sensitivity of 1ug/mL of FDP
  • No pretreatment of heparin specimen
  • No repeat testing due to false positives from heparin interference
Specifications
Specifications
Sample Collection Tubes
30 Tubes