Thermo Scientific™ Pacific Hemostasis™ FDP Assay Available on GSA/VA Contract for Federal Government customers only.
Measure fibrinogen degradation products (FDP) in serum and urine.
 View more versions of this product
Catalog No. 23-249991
  Check Availability
Add to Cart
    Specifications
    Description Latex Reagent
    Quantity 1 x 5mL,1 x 5mL
    Measure fibrinogen degradation products (FDP) in serum and urine with Thermo Scientific™ Pacific Hemostasis™ FDP Assay, a fast, easy-to-use latex agglutination assay used in the diagnosis of disseminated intravascular coagulation (DIC), deep vein thrombosis (DVT), pulmonary embolism (PE), certain pregnancy disorders and myocardial infarction.
    Features:
    • Latex sensitivity of 1ug/mL of FDP
    • No pretreatment of heparin specimen
    • No repeat testing due to false positives from heparin interference
  • Description & Specifications
Related Products
Product Content Feedback