Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls
Provide accurate, reproducible confirmation of coagulation test system performance with these coagulation controls.
 View more versions of this product
Catalog No. 23-029227
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Description INR Plasma Control, Levels 1-5
    Quantity 10 x 1mL,10 x 1mL
    Provide accurate, reproducible confirmation of coagulation test system performance with Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls. Controls can be run on most coagulation systems and are formulated to reflect expected patient results.
    Coagulation and Heparin control plasmas are prepared from fresh pools of human plasma.
    Coagulation Control Plasmas:
    • Normal control (Level 1) produces normal PT and aPTT times
    • Abnormal controls monitor therapeutic (Level 2) and highly elevated (Level 3) PT/aPTT times 
    Heparin Control Plasmas:
    • Simulate low (Level 1) and elevated (Level 2) plasma levels of Heparin
    • Use to monitor and evaluate patients on Heparin therapy
  • Description & Specifications
Related Products
Product Content Feedback