Learn More
Thermo Scientific™ Pacific Hemostasis™ Fibrinogen/Fibrin Analysis
Screen and quantify plasma fibrinogen based on the Clauss method.
 View more versions of this product
Catalog No. 23-176339
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This product is regulated by the federal government, and may not be ordered online.
Specifications
Description
Screen and quantify plasma fibrinogen based on the Clauss method with Thermo Scientific™ Pacific Hemostasis™ Fibrinogen/Fibrin Analysis.
Fibrinogen Assay Set:
  • Quantifies plasma fibrinogen levels
  • Detects acquired and congenital hemorrhagic disorders
  • Components available separately
Thrombin Time Assay:
  • Provides qualitative determination
  • Useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot
  • Reconstituted stability: 8 hours at 2° to 8°C
Ordering Information:
Products can be purchased as a set or individually.
Description & Specifications
Specifications
Thrombin Time Reagent
10 x 1mL
2° to 8°C