Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paclobutrazol, Alfa Aesar™

Click to view available options
Quantity:
5g
76738-62-0
C15H20ClN3O
293.795
MFCD01678673
RMOGWMIKYWRTKW-LSLKUGRBSA-N
^a-tert-Butyl-?-(4-chlorobenzyl)-1H-1,2,4-triazole-1-ethanol
53297383
(2S)-1-(4-chlorophenyl)-4,4-dimethyl-2-(1,2,4-triazol-1-yl)pentan-3-ol
CC(C)(C)C(C(CC1=CC=C(C=C1)Cl)N2C=NC=N2)O
76738-62-0
C15H20ClN3O
293.795
MFCD01678673
RMOGWMIKYWRTKW-LSLKUGRBSA-N
^a-tert-Butyl-?-(4-chlorobenzyl)-1H-1,2,4-triazole-1-ethanol
53297383
(2S)-1-(4-chlorophenyl)-4,4-dimethyl-2-(1,2,4-triazol-1-yl)pentan-3-ol
CC(C)(C)C(C(CC1=CC=C(C=C1)Cl)N2C=NC=N2)O
UN Number UN1325
Beilstein 6846840
Merck Index 14,6984
Formula Weight 293.79
Physical Form Powder
Melting Point 165°C to 166°C
Quantity 5g
Sensitivity Hygroscopic
Chemical Name or Material Paclobutrazol

GHS H Statement
H228-H302-H312
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P280-P240-P241-P301+P312-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : XZ4803300