Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PACRG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543934
Description
Peptide sequence: GAIMARCNLD HLGSSDPPTS ASQVAEIIVN SGDGIDYSQQ KRENIGDLIQ Sequence homology: Cow: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
PACRG is a protein that is conserved across metazoans. In vertebrates, this gene is linked in a head-to-head arrangement with the adjacent parkin gene, which is associated with autosomal recessive juvenile Parkinson's disease. These genes are co-regulated in various tissues and they share a bi-directional promoter. Both genes are associated with susceptibility to leprosy. The parkin co-regulated gene protein forms a large molecular complex with chaperones, including heat shock proteins 70 and 90, and chaperonin components. This protein is also a component of Lewy bodies in Parkinson's disease patients, and it suppresses unfolded Pael receptor-induced neuronal cell death.
Specifications
PACRG | |
Polyclonal | |
Unconjugated | |
PACRG | |
1700008H23Rik; FLJ32724; GLUP; HAK005771; Hypertension-related protein 1-like protein; molecular chaperone/chaperonin-binding protein; PACRG; PARK2 coregulated; PARK2 co-regulated; PARK2 coregulated gene protein; PARK2CRG; parkin coregulated; parkin coregulated gene protein; parkin co-regulated gene protein; Parkin coregulated gene protein homolog; RGD1561027; RP3-495O10.2; Unknown (protein for MGC:134579); zgc:101786 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
135138 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96M98 | |
PACRG | |
Synthetic peptide directed towards the middle region of human PACRG. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction