Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACRG Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | PACRG |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP154802
|
Novus Biologicals
NBP154802 |
100 μL |
Each of 1 for $436.00
|
|
Description
PACRG Polyclonal specifically detects PACRG in Human samples. It is validated for Western Blot.Specifications
PACRG | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ32724, Glup, HAK005771, Molecular chaperone/chaperonin-binding protein, PARK2 co-regulated, PARK2 coregulated gene protein, PARK2CRG, parkin coregulated gene protein, parkin co-regulated gene protein, RP3-495O10.2 | |
PACRG | |
IgG | |
Affinity Purified | |
33 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q96M98 | |
135138 | |
Synthetic peptides corresponding to PACRG(PARK2 co-regulated) The peptide sequence was selected from the middle region of PACRG. Peptide sequence GAIMARCNLDHLGSSDPPTSASQVAEIIVNSGDGIDYSQQKRENIGDLIQ. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title