Learn More
Abnova™ PACSIN2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00011252T01
Catalog No. 89-955-558
Add to cart
Specifications
Specifications
293T
BC008037.2
pCMV-PACSIN2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
PACSIN2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 53.57kDa
Transient overexpression cell lysate was tested with anti-PACSIN2 antibody (Mfr. No. H00011252-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human