Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACSIN3 Monoclonal Antibody (OTI2D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA502250
Description
This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.Specifications
PACSIN3 | |
Monoclonal | |
0.94 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UKS6 | |
PACSIN3 | |
Full length human recombit protein of human PACSIN3 produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
OTI2D7 | |
Unconjugated | |
PACSIN3 | |
SDPIII | |
Mouse | |
Affinity Chromatography | |
RUO | |
29763, 311187, 475984 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction