Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACSIN3 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: OTI4F8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273221FR
Description
PACSIN3 Monoclonal specifically detects PACSIN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
PACSIN3 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PACSIN3 | |
Full length recombinant protein of human PACSIN3 (NP_057307) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4F8 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Endophilin I, FLJ17861, protein kinase C and casein kinase substrate in neurons 3, protein kinase C and casein kinase substrate in neurons protein 3, SDPIII, SH3 domain-containing protein 6511, syndapin III | |
Mouse | |
Affinity Purified | |
Cancer, Diabetes Research, GPCR, Neuroscience, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
29763.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title