Learn More
PADI4 Mouse anti-Human, Clone: OTI4H5, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504813S
Catalog No. 50-167-5424
Add to cart
Description
Description
Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at  ft.Arg-8 ft. and/or  ft.Arg-17 ft. and histone H4 at  ft.Arg-3 ft., which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at  ft.Arg-2142 ft., which favors its interaction with NCOA2/GRIP1.
Specifications
Specifications
PADI4
Monoclonal
Unconjugated
PADI4
Liquid
PADI4
Full length human recombit protein of human PADI4 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4H5
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UM07
PAD, PAD4, PADI5, PDI4, PDI5
Mouse
IgG2a
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23569