Learn More
Brady™ Padlock Station w/ 5 Safety Padlocks
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Padlock Station is made of thermoformed polypropylene for rugged durabilty. Semi-transparent hinged cover with locking mechanism protects contents from harsh environments and prevents unauthorized access.
Manufacturer:  Brady™ 105928
Catalog No. 19-104-254
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Yellow
(1) padlock station; (5) kd safety padlocks, 1.5 in. shackle, red; (2) 1 in. lockout hasps; (2) 1.5 in. lockout hasps; (5) lockout tags
Thermoformed polypropylene
2.5 in. x 13.25 in. x 17 in.
Safety
17 in.
2.5 in.
13.25 in.