Learn More
Brady™ Long Body Keyed Alike Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks electrical current from passing padlock shackle to cylinder
Manufacturer:  Brady™ 123414
 View more versions of this product
Catalog No. 19-103-497
Add to cart
Description
Description
  • Lock with 7.6cm (3 in.) long body provides ample space for identification information
  • Made of nylon plastic
  • Lock labels are supplied with English and Spanish or English and French “DO NOT OPERATE” legends
  • Custom keying and engraving is available
  • Available in a variety of colors and packaging
Specifications
Specifications
3/Pk.
Red
Alike
Nylon plastic