Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAF1 Polyclonal Antibody, Active Motif™
Rabbit Polyclonal Antibody
$147.50 - $565.00
Specifications
Antigen | PAF1 |
---|---|
Concentration | Lot-Specific |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3505
|
Active Motif
61568 |
10 μL |
Each for $147.50
|
|
|||||
50-199-3504
|
Active Motif
61567 |
100 μL |
Each for $565.00
|
|
|||||
Description
This gene encodes a subunit of the polymerase associated factor (PAF1) complex. The PAF1 complex interacts with RNA polymerase II and plays a role in transcription elongation as well as histone modifications including ubiquitylation and methylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
PAF1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8N7H5 | |
54623 | |
This antibody was raised against a peptide within human PAF1. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Paf1 |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
5730511K23Rik; AA675045; F23149_1; hPAF1; Paf1; PAF1 homolog, Paf1/RNA polymerase II complex component; Paf1, RNA polymerase II associated factor, homolog; Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae); Pancreatic differentiation protein 2; PD2; RNA polymerase II-associated factor 1 homolog | |
Paf1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title