Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAF49 Mouse anti-Human, FITC, Clone: 635, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243633F

Catalog No. NBP243633F


Add to cart

Description

Description

PAF49 Monoclonal antibody specifically detects PAF49 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PAF49
635
Immunocytochemistry/Immunofluorescence
Purified
CD3EAP
Recombinant protein encompassing a sequence within the center region of human PAF49. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
A34.5, antisense to ERCC 1, Antisense to ERCC-1 protein, ASE-1ASE1, CASTMGC118851, CD3 epsilon associated protein, CD3e antigen, epsilon polypeptide associated protein, CD3e molecule, epsilon associated protein, CD3E-associated protein, CD3-epsilon-associated protein, DNA-directed RNA polymerase I subunit RPA34, PAF49RNA polymerase I-associated factor PAF49
Mouse
IgG2a
0.1 ml
Primary
10849
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit