Learn More
Aviva Systems Biology Corp PAFAH1B2 Antibody - N-terminal region (ARP58507_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58507_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAFAH1B2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAFAH1B2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62331