Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAFAH1B3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179625

 View more versions of this product

Catalog No. NBP179625


Add to cart

Description

Description

PAFAH1B3 Polyclonal antibody specifically detects PAFAH1B3 in Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

PAFAH1B3
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
PAFAH1B3
Synthetic peptide directed towards the middle region of human PAFAH1B3The immunogen for this antibody is PAFAH1B3. Peptide sequence GHTAEQVTGGIKAIVQLVNERQPQARVVVLGLLPRGQHPNPLREKNRQVN.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Mouse: 100%; Canine: 92%; Guinea pig: 92%.
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat
Western Blot
Western Blot 1:1000
NP_002564
EC 3.1.1.47, FLJ44990, PAF acetylhydrolase 29 kDa subunit, PAF-AH 29 kDa subunit, PAFAH subunit gamma, PAF-AH subunit gamma, PAF-AH1b alpha 1 subunit, PAFAHG, platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa), platelet-activating factor acetylhydrolase IB subunit gamma, platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit (29kD), platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa)
Rabbit
IgG
100 ul
Neuroscience
Primary
5050
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit