Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAGE5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550470

Catalog No. PA550470


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total PAGE5 protein.

This gene is a member of family of proteins that are expressed in a variety of tumors and in some fetal and reproductive tissues. The encoded protein may protect cells from programmed cell death. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found. [provided by RefSeq, Jan 2015].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAGE5
Polyclonal
Unconjugated
PAGE5
cancer/testis antigen 16.1; cancer/testis antigen family 16, member 1; cancer/testis antigen family 16, member 2; CT16; CT16.1; CT16.2; CT161; G antigen family E member 1; GAGEE1; P antigen family member 5; P antigen family, member 5 (prostate associated); PAGE family member 5; PAGE5; PAGE-5; Prostate-associated gene 5 protein
Rabbit
Antigen affinity chromatography
RUO
90737
-20°C
Liquid
Immunohistochemistry (Paraffin)
2.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q96GU1
PAGE5
Full length fusion protein of Human PAGE5.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.