Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAGE5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548452

Catalog No. PA548452

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is a member of family of proteins that are expressed in a variety of tumors and in some fetal and reproductive tissues. The encoded protein may protect cells from programmed cell death. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found. [provided by RefSeq, Jan 2015]
Specifications

Specifications

PAGE5
Polyclonal
PBS with 0.09% sodium azide
Q96GU1
P antigen family member 5, PAGE-5, Cancer/testis antigen 161, CT161, G antigen family E member 1, Prostate-associated gene 5 protein, PAGE5, GAGEE1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90737
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PAGE5
Liquid
PAGE5
KLH conjugated synthetic peptide between 102-130 amino acids from the C-terminal region of human PAGE5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit