Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB2 Mouse anti-Human, Clone: OTI1E3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525776

Catalog No. PIMA525776


Add to cart

Description

Description

PAI2 inhibits urokinase-type plasminogen activator. The monocyte derived PAI-2 is distinct from the endothelial cell-derived PAI-1.
Specifications

Specifications

SERPINB2
Monoclonal
0.53 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P05120
HsT1201, PAI, PAI-2, PAI2, PLANH2, monocyte Arg-serpin, placental plasminogen activator inhibitor, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, serpin B2, urokinase inhibit
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5055
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1E3
Unconjugated
SERPINB2
Liquid
SERPINB2
Full length human recombit protein of SERPINB2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit