Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAIP1 Polyclonal Antibody
PAIP1 Polyclonal Antibody
Supplier: Thermo Scientific PA530394
Description
PAIP1 Polyclonal Antibody for Western Blot

Specifications
PAIP1 | |
Polyclonal | |
Unconjugated | |
PAIP1 | |
PABC1-interacting protein 1; PABP-interacting protein 1; Paip1; PAIP-1; poly(A) binding protein interacting protein 1; poly(A)-binding protein-interacting protein 1; polyadenylate binding protein-interacting protein 1; polyadenylate-binding protein-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10605 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.79 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H074 | |
PAIP1 | |
Recombinant fragment corresponding to a region within amino acids 178 and 443 of Human PAIP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction