Learn More
PAK1IP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160711AP
Catalog No. 50-173-0691
Add to cart
Description
Description
Proteins that directly bind to nascent RNA polymerase II transcripts, the heterogenous nuclear ribonucleoproteins (hnRNPs), play an important role in both transcript-specific packaging and alternative splicing of pre-mRNAs. A group of abundant hnRNPs, the M1-M4 proteins, appear as a cluster of four proteins. The M proteins are pre-mRNA binding proteins in vivo, and they bind avidly to poly (G) and poly (U) RNA homopolymers in vitro. M proteins are members of the ribonucleoprotein consensus sequence family of RNA-binding proteins with greatest similarity to a hypothetical RNA-binding protein from Saccharomyces cerevisiae. The M proteins also possess an unusual hexapeptide-repeat region rich in methionine and arginine residues (MR repeat motif) that resembles a repeat in the 64kDa subunit of cleavage stimulation factor, which is involved in 3 ft.-end maturation of pre-mRNAs. Proteins immunologically related to M exist in divergent eukaryotes ranging from human to yeast.
Specifications
Specifications
PAK1IP1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9DCE5, Q9NWT1
hPIP1, MAK11, PAK/PLC interacting protein 1, PAK1 interacting protein 1, PAK1IP1, PIP1, RP11 421M1.5, WDR84
Rabbit
IgG
150 μL
-20°C
Primary
361232, 55003, 68083
Western Blot
Unconjugated
PAK1IP1
Liquid
Pak1ip1
PAK1IP1 Fusion Protein Ag9016
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat