Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK4 Rabbit anti-Human, Mouse, Rat, Hamster, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$176.85 - $436.00
Specifications
Antigen | PAK4 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 4 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
PAK4 Polyclonal specifically detects PAK4 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot.Specifications
PAK4 | |
Western Blot 0.5 - 4 ug/mL | |
Polyclonal | |
Rabbit | |
Apoptosis, Protein Kinase | |
BAF84528 | |
10298 | |
Synthetic peptide corresponding to the N-terminus of human Pak4 (BAF84528). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Hamster | |
EC 2.7.11, EC 2.7.11.1, KIAA1142, p21 protein (Cdc42/Rac)-activated kinase 4, p21(CDKN1A)-activated kinase 4, p21-activated kinase 4, PAK-4, protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs, serine/threonine-protein kinase PAK 4 | |
PAK4 | |
IgG | |
Affinity Purified | |
The antibody recognizes ∼60-70 kDa Pak4 in samples of human, mouse, and rat origins. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title