Learn More
Abnova™ PAK5 (Human) Recombinant Protein
For use with Functional Study.
Manufacturer:  Abnova™ P6534
Catalog No. 89-339-423
Add to cart
Description
Description
Human PAK5 (NP_065074.1, 425 a.a. - 719 a.a.) partial recombinant protein with GST-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
Functional Study
p21 protein (Cdc42/Rac)-activated kinase 7
5 µg
KIAA1264, MGC26232, PAK5
57144
The purity was assessed by SDS-PAGE/CBB staining.
Stored at -80°C. Aliquot to avoid repeated freezing and thawing.