Learn More
Thermco™ PAKSENSE™ Temp and Humidity Cold Chain Data Logger
Thermco™ PAKSENSE™ Temp and Humidity Cold Chain Data Logger record temperature and humidity levels of a product's environment during distribution and storage
Manufacturer:  Thermco™ XPDHURED
Catalog No. 04-500-048
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Both temperature and humidity ranges are programmable via the PakSense TH Software
  • All units are reusable
  • Optimal for use in closed-loop situations and import/export container shipments.
  • Humidity and temperature data can be downloaded and graphed
  • All data collected by the data loggers can be saved electronically and is easily printed and shared.
Specifications
Specifications
1 minute sampling
No
±1.0°C
15 month shelf life
-40 to 85°C and 0% to 100%Rh