Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palatine Fast Yellow BLN, pure, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
10127-05-6
C18H14N4Na2O9S2
540.429
MFCD07189471
LQKWRCLAYBSICM-PFNMRQQASA-L
Acid Yellow 54,C.I. 19010
92043787
disodium;4-carboxy-3-[(2Z)-2-[3-methyl-1-(2-methyl-4-sulfonatophenyl)-5-oxopyrazol-4-ylidene]hydrazinyl]benzenesulfonate
CC1=C(C=CC(=C1)S(=O)(=O)[O-])N2C(=O)C(=NNC3=C(C=CC(=C3)S(=O)(=O)[O-])C(=O)O)C(=N2)C.[Na+].[Na+]
10127-05-6
C18H14N4Na2O9S2
540.429
MFCD07189471
LQKWRCLAYBSICM-PFNMRQQASA-L
Acid Yellow 54,C.I. 19010
92043787
disodium;4-carboxy-3-[(2Z)-2-[3-methyl-1-(2-methyl-4-sulfonatophenyl)-5-oxopyrazol-4-ylidene]hydrazinyl]benzenesulfonate
CC1=C(C=CC(=C1)S(=O)(=O)[O-])N2C(=O)C(=NNC3=C(C=CC(=C3)S(=O)(=O)[O-])C(=O)O)C(=N2)C.[Na+].[Na+]
Infrared Spectrum Authentic
Formula Weight 562.4
Physical Form Powder
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Brown-Rust
Quantity 25g
Chemical Name or Material Palatine Fast Yellow BLN