Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PALDA, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
136181-87-8
C24H41NO3
391.596
TWJJFOWLTIEYFO-UHFFFAOYSA-N
palda, palmitoyl dopamine, n-palmitoyldopamine, n-palmitoyl dopamine, n-2-3,4-dihydroxyphenyl ethyl hexadecanamide, bml3-g01
16759163
N-[2-(3,4-dihydroxyphenyl)ethyl]hexadecanamide
CCCCCCCCCCCCCCCC(=O)NCCC1=CC(=C(C=C1)O)O
This item is not returnable. View return policy
136181-87-8
C24H41NO3
391.596
TWJJFOWLTIEYFO-UHFFFAOYSA-N
palda, palmitoyl dopamine, n-palmitoyldopamine, n-palmitoyl dopamine, n-2-3,4-dihydroxyphenyl ethyl hexadecanamide, bml3-g01
16759163
N-[2-(3,4-dihydroxyphenyl)ethyl]hexadecanamide
CCCCCCCCCCCCCCCC(=O)NCCC1=CC(=C(C=C1)O)O

Endogenous lipid; potentiates effects of endovanilloids at TRPV1 receptors

Endogenous fatty acid dopamide that displays 'entourage' effects on endovanilloids NADA and anandamide. Inactive at TRPV1 and CB1 receptors (at concentrations up to 5 μM) and does not inhibit AMT or FAAH (IC50 > 25 μM). However, potentiates TRPV1-mediated effects of NADA; lowers EC50 from ∽ 90 to ∽ 30 nM.
Quantity 10mg
Formula Weight 391.59
Percent Purity >98%
Chemical Name or Material PALDA

Recommended Storage : Store at +4°C